

Lithium hexafluoroarsenate
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:29935-35-1
:
Formula:AsF₆Li
MDL Number:MFCD00011076
:195.85 g/mol
Melting Pt:349 °C
EINECS:249-963-0
UN:1557
ADR:6.1,II
:GHS06
:GHS09
Frequently Bought Together