Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
1-Ethynylcyclohexanol

1-Ethynylcyclohexanol

Glass bottle for solids, 100 g
Catalog # MOLEM86812236
Flammable
Irritant
Marketsource
Supplier:   Molekula
1-Ethynylcyclohexanol
Catalog # MOLEM86812236
Supplier:  Molekula
Marketsource
CAS Number:  78-27-3
1-Ethynylcyclohexanol

Glass bottle for solids, 100 g

CAS Number:  78-27-3
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Ethynyl-1-cyclohexanol
  • CAS Number:78-27-3
  • Formula:C₈H₁₂O
  • MDL Number:MFCD00003858
  • Molecular Weight:124.18 g/mol
  • Boiling Pt:161 °C (1013 hPa)
  • Melting Pt:20 °C
  • Density:0.9737 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:241-385-7
  • UN:1325
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications