

Vitamin A palmitate, Supelco®
Catalog # 46959-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:2440
- Pk Info:100 mg
Specifications
About this item
:
:524.87 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-228-5
:GHS08
:GHS07
CAS Number:79-81-2
Formula:C₃₆H₆₀O₂
MDL Number:MFCD00019414