3-Nitropropionic acid, Sigma-Aldrich®
Catalog # SIALN22908-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:119.08 g/mol
:M-Clarity™ quality level = MQ100
EINECS:208-003-0
CAS Number:504-88-1
Formula:C₃H₅NO₄
MDL Number:MFCD00007406