Methyl-2,4-difluorobenzoate, Sigma-Aldrich®
Catalog # SIAL595047-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:172.13 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:106614-28-2
Formula:C₈H₆F₂O₂
MDL Number:MFCD03093799