Methyl-2,4-difluorobenzoate ≥98%
Catalog # H32161.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
CAS Number:106614-28-2
:
Formula:C₈H₆F₂O₂
MDL Number:MFCD03093799
:172.13 g/mol
Storage Temperature:Ambient