Order Entry
United Kingdom
ContactUsLinkComponent
 
Diethyl terephthalate 98%
 
undefined
Diethyl terephthalate 98%

 

  • Appearance:
    White to cream Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    43 to 47 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together