Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Cyclohexanecarboxylic acid ≥98%

Cyclohexanecarboxylic acid ≥98%

Irritant
Marketsource
Cyclohexanecarboxylic acid ≥98%
Marketsource
Certificates
Cyclohexanecarboxylic acid ≥98%
Cyclohexanecarboxylic acid ≥98%

4 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 98-89-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 118°(244°F) Refractive Index: 1.4610 Melting Point: 29-32° Boiling Point: 231-233° Beilstein Reference: 970529 Merck Reference: 14,2724 MDL No.: MFCD00001461

Synonyms:  Hexahydrobenzoic acid
  • CAS Number:98-89-5
  • Formula:C₇H₁₂O₂
  • MDL Number:MFCD00001461
  • Molecular Weight:128.17 g/mol
  • Merck Index:14,2724
  • Boiling Pt:231…233 °C
  • Melting Pt:29…32 °C
  • Density:1.048
  • Flash Pt:118 °C (244 °F)
  • Storage Temperature:Ambient
  • EINECS:202-711-3
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.