2-Phenylbenzoic acid 98%
: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid
- :947-84-2
- :C₁₃H₁₀O₂
- :MFCD00002463
- :198.22 g/mol
- :199 °C (10 mmHg)
- :110…114 °C
- :1,458 g/cm³ (20 °C)
- :Ambient
- :213-432-1
: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid
- :947-84-2
- :C₁₃H₁₀O₂
- :MFCD00002463
- :198.22 g/mol
- :199 °C (10 mmHg)
- :110…114 °C
- :1,458 g/cm³ (20 °C)
- :Ambient
- :213-432-1
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:110 to 114 °C
- HPLC:≥97.5 %
- Titration with NaOH:≥97.5 %
Product Family Options
- Pack typePk



