p-Xylene, Supelco®
Glass bottle, 50 mL
Catalog # 95680-50ML
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: 1,4-Dimethylbenzene
- CAS Number:106-42-3
- Formula:C₆H₄(CH₃)₂
- MDL Number:MFCD00008556
- Molecular Weight:106.17 g/mol
- Merck Index:12,10214
- Boiling Pt:138.3 °C (1013 hPa)
- Melting Pt:13 °C
- Density:0.861 g/cm³ (20 °C)
- Flash Pt:25 °C
- Storage Temperature:Ambient
- UN:1307
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- EINECS:203-396-5
Specifications
Specifications
- Pack type:Glass bottle
- Pk:50 mL



