Benzo[b]thiophene-3-boronic acid, Sigma-Aldrich®
Catalog # SIAL512117-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.02 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:113893-08-6
Formula:C₈H₇BO₂S
MDL Number:MFCD01114669