Order Entry
United Kingdom
ContactUsLinkComponent
Benzoin, Sigma-Aldrich®
  SIAL399396-5G
 :  Merck
 :  
Benzoin, Sigma-Aldrich®
  SIAL399396-5G
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
 :
 :212.25 g/mol
 :M-Clarity™ quality level = MQ100
 :204-331-3
 :119-53-9
 :C₁₄H₁₂O₂
 :MFCD00004496