Order Entry
United Kingdom
ContactUsLinkComponent
 
Organic reference standard, Naphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20905000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Naphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20905000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    91-20-3
  • Description:
    Naphthalene 10 µg/ml in acetonitrile
  • Formula:
    C₁₀H₈
  • Description:
    Naphthalene

Specifications

About this item