Order Entry
United Kingdom
ContactUsLinkComponent
 
Organic reference standard, Acenaphthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20505000AL
 :  
Organic reference standard, Acenaphthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20505000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    83-32-9
  • Description:
    Acenaphthene 10 µg/ml in acetonitrile
  • Formula:
    C₁₂H₁₀
  • Description:
    Acenaphthene