Organic reference standard, 2,6-Dimethylnaphthalene 50 µg/ml in toluene, CIL
Catalog # PROOCILULM72711.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:581-42-0
- Description:2,6-Dimethylnaphthalene 50 µg/ml in toluene
- Formula:C₁₂H₁₂
- Description:2,6-Dimethylnaphthalene