tert-Butyl nitrite 90%, pure
:540-80-7
:
:C₄H₉NO₂
:MFCD00002055
:103.12 g/mol
:61…63 °C (760 mmHg)
:0.86 g/cm³
:–10 °C
:Refrigerator
:208-757-0
:2351
:3,II
:GHS07
:GHS02
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3670 to 1.3700 (20°C, 589 nm)
- GC:≥88 %
Frequently Bought Together