(+)-Valencene
Glass bottle for solids, 5 g
ABCRAB168272
Marketsource
: abcr
: Valencene (Standard), (3R,4aS,5R)-4a,5-Dimethyl-3-(prop-1-en-2-yl)-1,2,3,4,4a,5,6,7-octahydronaphthalene
- :4630-07-3
- :C₁₅H₂₄
- :MFCD00075884
- :204.36 g/mol
- :225-047-6
- Pack type:Glass bottle for solids
- Pk:5 g