Boc-Pro-OH
Catalog # ABCRAB138550.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:215.25 g/mol
EINECS:239-848-3
CAS Number:15761-39-4
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD00037324