
4-Hydroxy-3,5-dimethylbenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2233-18-3
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00006946
:150.18 g/mol
Melting Pt:112…116 °C
EINECS:218-774-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together