Order Entry
United Kingdom
ContactUsLinkComponent
sec-Butylbenzene 99+%
Flammable
sec-Butylbenzene 99+%
Catalog # ACRO107860050
CAS Number:  
undefined
Flammable
sec-Butylbenzene 99+%
Catalog # ACRO107860050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4890 to 1.4910 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

About this item

 :
 :134.22 g/mol
Merck Index:13,01549
Boiling Pt:173 °C (1013 hPa)
Melting Pt:–75 °C
Density:0.86 g/cm³
Flash Pt:45 °C
Storage Temperature:Ambient
EINECS:205-227-0
UN:2709
ADR:3,III
 :GHS02
CAS Number:135-98-8
Formula:C₆H₅CH(CH₃)CH₂CH₃
MDL Number:MFCD00009329