
2-Methylanisole 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:578-58-5
:
Formula:C₈H₁₀O
MDL Number:MFCD00008373
:122.17 g/mol
Boiling Pt:172 °C (1013 hPa)
Density:0.98 g/cm³
Flash Pt:51 °C
Storage Temperature:Ambient
EINECS:209-426-3
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5180 (20°C, 589 nm)
- GC:≥98.5 %
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0005 meq/g
- Free chlorine:≤0.00003 % (as Cl)
- Aluminium (Al):≤0.5 ppm
- Barium (Ba):≤0.1 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Tin (Sn):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Aldehyde (HCHO):≤0.0005 %
- Ethanol:≤0.3 % (GC)
Specification Test Results
Frequently Bought Together