Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α,α,4-Tetrachlorotoluene
α,α,α,4-Tetrachlorotoluene
α,α,α,4-Tetrachlorotoluene
EHERC17380000
:
EHRENSTORFER
:
α,α,α,4-Tetrachlorotoluene
EHERC17380000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
229.92 g/mol
:
245 °C (1013 hPa)
:
5.8 °C
:
1.5036 g/cm³ (20 °C)
:
226-009-1
:
3265
:
8,II
:
5216-25-1
:
C₇H₄Cl₄
:
MFCD00000790