
3-Methyl-4-nitrobenzoic acid 99%
:3113-71-1
:
:C₈H₇NO₄
:MFCD00007168
:181.15 g/mol
:356 °C (1013 hPa)
:216…219 °C
:Ambient
:221-479-4
:2811
:6.1,III
:GHS07
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Melting point:216 to 219 °C
- Titration with NaOH:≥98.5 %
Frequently Bought Together