Order Entry
United Kingdom
ContactUsLinkComponent
 
4-Tolualdehyde 99+%
 
undefined
4-Tolualdehyde 99+%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5440 to 1.5480 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Specific gravity:
    (20°C) 1.010 to 1.025

 

 

Frequently Bought Together