Order Entry
United Kingdom
ContactUsLinkComponent
 
1-Chloro-2-butene (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL28115-100ML
Supplier:  Merck
CAS Number:  
1-Chloro-2-butene (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL28115-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :90.55 g/mol
Merck Index:12,02181
Boiling Pt:83…84 °C (1013 hPa)
Density:0.9491 g/cm³ (0 °C)
EINECS:209-739-5
CAS Number:591-97-9
Formula:C₄H₇Cl
MDL Number:MFCD00000987