Dichloromethane
205 L
Catalog # BARRMECH180
Marketsource
Supplier: BARRETTINE INDUSTRIAL
Dichloromethane
Catalog # BARRMECH180
Supplier: BARRETTINE INDUSTRIAL
Marketsource
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39.8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1.322 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
Specifications
Specifications
- Pk:205 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



