Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Phenethyl bromide

Phenethyl bromide

Irritant
Marketsource
Supplier:   FLUOROCHEM
Phenethyl bromide
Supplier:  FLUOROCHEM
Marketsource
Certificates
Phenethyl bromide

2 Options Available Below

Product Details & Documents
Synonyms:  (2-Bromoethyl)benzene, 2-Phenylethyl bromide
  • CAS Number:103-63-9
  • Formula:BrCH₂CH₂C₆H₅
  • MDL Number:MFCD00000240
  • Molecular Weight:185.06 g/mol
  • Boiling Pt:221 °C (1013 hPa)
  • Melting Pt:–56 °C
  • Density:1,37 g/cm³ (20 °C)
  • Flash Pt:89 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-130-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.