Order Entry
United Kingdom
ContactUsLinkComponent
4-Vinylbenzoic acid
  MOLE51779787-1G
 :  Molekula
 :  
4-Vinylbenzoic acid
  MOLE51779787-1G
 :  Molekula
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
 :
 :148.16 g/mol
 :282 °C (1013 hPa)
 :282 °C (1013 hPa)
 :115 °C
 :1,3 g/cm³ (20 °C)
 :Refrigerator
 :214-053-4
 :GHS07
 :1075-49-6
 :C₉H₈O₂
 :MFCD00002569