Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
5-Bromo-2-methylanisole ≥97%

5-Bromo-2-methylanisole ≥97%

5-Bromo-2-methylanisole ≥97%
Certificates
5-Bromo-2-methylanisole ≥97%
5-Bromo-2-methylanisole ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 67868-73-9 Molecular Formula: C8H9BrO Formula Weight: 201.06 Refractive Index: 1.5630 Boiling Point: 102-104°/13mm MDL No.: MFCD06797972

Synonyms:  4-Bromo-2-methoxy-1-methylbenzene
  • CAS Number:67868-73-9
  • Formula:C₈H₉BrO
  • MDL Number:MFCD06797972
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:102…104 °C (13 mmHg)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £30.90
    £30.90
    1 * 1 g
       £30.90

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £96.00
    £96.00
    1 * 5 g
       £96.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.