Phenol, crystals, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Boiling Pt:181.8 °C (1013 hPa)
- Melting Pt:40.8 °C
- Density:1.06 g/cm³ (20 °C)
- Flash Pt:81 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:203-632-7
- ADR:6.1,II
- :M-Clarity™ quality level = MQ200
- UN:1671
4 Options Available Below
Product Details & Documents
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Boiling Pt:181.8 °C (1013 hPa)
- Melting Pt:40.8 °C
- Density:1.06 g/cm³ (20 °C)
- Flash Pt:81 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:203-632-7
- ADR:6.1,II
- :M-Clarity™ quality level = MQ200
- UN:1671
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



