Diethyl dibromomalonate, Sigma-Aldrich®
Catalog # SIAL412708-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:317.96 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:211-151-9
UN:3265
CAS Number:631-22-1
Formula:C₇H₁₀Br₂O₄
MDL Number:MFCD00015154