1-Ethyl-3-methylimidazolium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1-Ethyl-3-methylimidazolium Hydrogen sulphate
CAS Number:412009-61-1
:
Formula:C₆H₁₂N₂O₄S
MDL Number:MFCD06798195
Molecular Weight:208.24 g/mol
UN:3265
ADR:8 II
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together