
1-Butyl-3-methylimidazolium methanesulphonate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:342789-81-5
:
Formula:C₉H₁₈N₂O₃S
MDL Number:MFCD06798173
:234.32 g/mol
Melting Pt:75…80 °C
Flash Pt:119 °C (246 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together