
Diethyl ethoxymethylenemalonate 99+%
:87-13-8
:
:C₁₀H₁₆O₅
:MFCD00009148
:216.23 g/mol
:1.08 g/cm³
:144 °C
:201-725-7
:GHS08
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4610 to 1.4630 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤20 APHA
Frequently Bought Together