Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Chloroform, Sigma-Aldrich®

Chloroform, Sigma-Aldrich®

5 Vial
Catalog # SIALC7559-5VL
Toxic
Health
Supplier:   Merck
Chloroform, Sigma-Aldrich®
Catalog # SIALC7559-5VL
Supplier:  Merck
CAS Number:  67-66-3
Chloroform, Sigma-Aldrich®
Chloroform, Sigma-Aldrich®

5 Vial

CAS Number:  67-66-3
Price
£169.00
Log in for availability
1 * 5 Vial
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-663-8
  • UN:1888
  • ADR:6.1,III
  • REACH:05-2114614906-45
Specifications

Specifications

  • Pk:
    5 Vial

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.