Order Entry
United Kingdom
ContactUsLinkComponent
Octamethylcyclotetrasiloxane ≥97%
Octamethylcyclotetrasiloxane ≥97%
  ABCRAB111277.2000
 :  abcr
 :  
Octamethylcyclotetrasiloxane ≥97%
  ABCRAB111277.2000
 :  abcr
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    2 kg
 :
 :296.62 g/mol
 :12,06843
 :175 °C (1013 hPa)
 :18 °C
 :0.95 g/cm³ (20 °C)
 :51 °C
 :209-136-7
 :1993
 :3,III
 :GHS08
 :556-67-2
 :[-Si(CH₃)₂O-]₄
 :MFCD00003269