2-Iodophenol
Glass bottle for solids, 100 g
MOLE14862771-100G
Marketsource
: Molekula
- :533-58-4
- :C₆H₅IO
- :MFCD00013963
- :220.01 g/mol
- :13,05057
- :186…187 °C (213 hPa)
- :39…42 °C
- :1.8757 g/cm³ (80 °C)
- :Ambient
- :208-569-9
- :2811
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:100 g