2-Iodophenol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 533-58-4 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 39-41° Boiling Point: 186-187°/160mm Beilstein Reference: 1855300 Merck Reference: 14,5035 MDL No.: MFCD00013963
- CAS Number:533-58-4
- Formula:C₆H₅IO
- MDL Number:MFCD00013963
- Molecular Weight:220.01 g/mol
- Merck Index:14,5035
- Boiling Pt:186…187 °C (160 mmHg)
- Melting Pt:39…41 °C
- Density:1.947
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:208-569-9
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 533-58-4 Molecular Formula: C6H5IO Formula Weight: 220.01 Flash Point: 110°(230°F) Melting Point: 39-41° Boiling Point: 186-187°/160mm Beilstein Reference: 1855300 Merck Reference: 14,5035 MDL No.: MFCD00013963
- CAS Number:533-58-4
- Formula:C₆H₅IO
- MDL Number:MFCD00013963
- Molecular Weight:220.01 g/mol
- Merck Index:14,5035
- Boiling Pt:186…187 °C (160 mmHg)
- Melting Pt:39…41 °C
- Density:1.947
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:208-569-9
- UN:2811
- ADR:6.1,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



