Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Iodophenol 98%

2-Iodophenol 98%

Irritant
2-Iodophenol 98%
Certificates
2-Iodophenol 98%

3 Options Available Below

Product Details & Documents
  • CAS Number:533-58-4
  • Formula:C₆H₅IO
  • MDL Number:MFCD00013963
  • Molecular Weight:220.01 g/mol
  • Merck Index:13,05057
  • Boiling Pt:186…187 °C (160 mmHg)
  • Melting Pt:37…43 °C
  • Density:1,8757 g/cm³ (80 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:208-569-9
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to brown-red to brown Crystalline powder or low melting mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    37 to 43 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £23.00
    £23.00
    1 * 5 g
       £23.00

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £58.80
    £58.80
    1 * 25 g
       £58.80

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £223.00
    £223.00
    1 * 100 g
       £223.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.