Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Fluorophenol ≥98%

2-Fluorophenol ≥98%

Toxic
Corrosive
Flammable
2-Fluorophenol ≥98%
Certificates
2-Fluorophenol ≥98%
2-Fluorophenol ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 367-12-4 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 46°(114°F) Refractive Index: 1.5140 Odour: Phenol-like Melting Point: 14-16° Boiling Point: 150-152° Beilstein Reference: 1905112 UN No.: 2924 MDL No.: MFCD00002155

Synonyms:  o-Fluorophenol
  • CAS Number:367-12-4
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002155
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:150…152 °C
  • Melting Pt:14…16 °C
  • Density:1.215
  • Flash Pt:46 °C (114 °F)
  • Storage Temperature:Ambient
  • EINECS:206-681-2
  • UN:2924
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.