1-Chloro-4-nitrobenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-00-5
:
Formula:C₆H₄ClNO₂
MDL Number:MFCD00007285
Merck Index:13,02170
Boiling Pt:239 °C (1013 hPa)
Melting Pt:81…84 °C
Density:1.298 g/cm³
Flash Pt:127 °C
Storage Temperature:Ambient
UN:1578
ADR:6.1,II
:157.56 g/mol
EINECS:202-809-6
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Yellow Crystals or flakes
- Infrared spectrum:Conforms
- Melting point:81 to 84 °C
- GC:≥98.5 %
- Titration with HClO4:98.5 to 101.5 % (on dried substance)
Specification Test Results
Frequently Bought Together