Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2,5-Dichlorobenzaldehyde 98%

2,5-Dichlorobenzaldehyde 98%

Corrosive
2,5-Dichlorobenzaldehyde 98%
Certificates
2,5-Dichlorobenzaldehyde 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:6361-23-5
  • Formula:C₇H₄Cl₂O
  • MDL Number:MFCD00156140
  • Molecular Weight:175.01 g/mol
  • Melting Pt:54…58 °C
  • EINECS:228-832-1
  • UN:3261

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    54 to 58 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £23.40
    £23.40
    1 * 1 g
       £23.40

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £76.50
    £76.50
    1 * 5 g
       £76.50

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.