Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2,3-Dichlorobenzaldehyde 99%

Corrosive
2,3-Dichlorobenzaldehyde 99%
Certificates
2,3-Dichlorobenzaldehyde 99%

2 Options Available Below

Product Details & Documents
  • CAS Number:6334-18-5
  • Formula:C₇H₄Cl₂O
  • MDL Number:MFCD00010127
  • Molecular Weight:175.01 g/mol
  • Boiling Pt:144 °C (30 mmHg)
  • Melting Pt:60…64 °C
  • Density:1,33 g/cm³ (100 °C)
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:228-711-3
  • UN:3261
  • ADR:8,II

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 64 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £40.60
    £40.60
    1 * 25 g
       £40.60

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £115.00
    £115.00
    1 * 100 g
       £115.00