Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Phenyl benzoate 99%

Phenyl benzoate 99%

Irritant
Phenyl benzoate 99%
Certificates
Phenyl benzoate 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Benzoic acid phenyl ester
  • CAS Number:93-99-2
  • Formula:C₁₃H₁₀O₂
  • MDL Number:MFCD00003072
  • Molecular Weight:198.22 g/mol
  • Merck Index:12,07429
  • Boiling Pt:299 °C (1013 hPa)
  • Melting Pt:68…70 °C
  • Density:1,041 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:202-293-2

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 70 °C
  • GC:
    ≥98.5 %
  • Acid value:
    ≤1 mg KOH/g
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £17.70
    £17.70
    1 * 5 g
       £17.70

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £36.80
    £36.80
    1 * 100 g
       £36.80

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £155.00
    £155.00
    1 * 500 g
       £155.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.