
Gallic acid 98%
:149-91-7
:
:C₇H₆O₅
:MFCD00002510
:170.12 g/mol
:13,04366
:501 °C (1013 hPa)
:253 °C (dec.)
:1.694 g/cm³
:205-749-9
:GHS07
:Ambient
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Frequently Bought Together