4-Hydroxy-3-nitrobenzoic acid for synthesis
8.40004.0025
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
:
:183.12 g/mol
:182…185 °C
:1 g/cm³ (20 °C)
:210-494-1
:GHS07
:Ambient
:616-82-0
:C₇H₅NO₅
:MFCD00007119