Sodium benzoate 99.0-100.5% Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Benzoic acid sodium salt
CAS Number:532-32-1
:
Formula:C₆H₅COONa
MDL Number:MFCD00012463
Density:1.44 g/cm³ (20 °C)
Molecular Weight:144.11 g/mol
Melting Pt:410…430 °C
Flash Pt:>100 °C
Merck Index:13,08654
EINECS:208-534-8
REACH:01-2119460683-35
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (calculated on dried substance):99.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- Acidity or alkalinity:Passes test
- Halogenated compounds:Passes test
- Heavy metals (as Pb):Max. 10 ppm
- Loss on drying (105°C):Max. 2 %
- Statement of original manufacturer:(*)
- Benzene (*):Max. 2 ppm
- Toluene (*):Max. 890 ppm
- Other residual solvents (*):Unlikely by manuf.process
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
83551_SODIUM BENZOATE PH.EUR._BSE_Rev1
BSE/TSE Certificates
spec_VWRC83551.290 spec_VWRC83551.290 spec_VWRC83551.360 spec_VWRC83551.360 spec_VWRC83551.461
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together