
Lithium benzoate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:553-54-8
:
Formula:C₇H₅LiO₂
MDL Number:MFCD00035540
:128.06 g/mol
Merck Index:14,5523
Melting Pt:>300 °C
Density:1.397 g/cm³ (20 °C)
EINECS:209-042-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together