
4-Cyanobenzoic acid
Catalog # MOLE21590404-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:147.13 g/mol
Boiling Pt:313 °C (1013 hPa)
Melting Pt:220…222 °C
Density:1,382 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-606-9
:GHS07
CAS Number:619-65-8
Formula:C₈H₅NO₂
MDL Number:MFCD00002528