Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Nitrosalicylaldehyde
5-Nitrosalicylaldehyde
5-Nitrosalicylaldehyde
MOLEM56349196
:
Molekula
:
5-Nitrosalicylaldehyde
MOLEM56349196
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
167.12 g/mol
:
123…126 °C
:
202-587-0
:
GHS07
:
Ambient
:
97-51-8
:
C₇H₅NO₄
:
MFCD00007337